N,N-Diallyl-5-(aminomethyl)-2-pyridinamine

Supplier: Matrix Scientific

052054-500MG 052054-2.5G
101898-106EA 364.62 USD
101898-106 101898-108
N,N-Diallyl-5-(aminomethyl)-2-pyridinamine
N,N-Diallyl-5-(aminomethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₇N₃
MDL Number: MFCD13561883
CAS Number: 1220030-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR