2-[[5-(Aminomethyl)-2-pyridinyl](ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

052053-500MG 052053-2.5G
101898-102EA 364.62 USD
101898-102 101898-104
2-[[5-(Aminomethyl)-2-pyridinyl](ethyl)amino]-1-ethanol
2-[[5-(Aminomethyl)-2-pyridinyl](ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₇N₃O
MDL Number: MFCD12813598
CAS Number: 1183030-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR