1-[5-(Aminomethyl)-2-pyridinyl]-3-pyrrolidinol dihydrochloride

Supplier: Matrix Scientific

052051-500MG 052051-2.5G
101898-094EA 364.62 USD
101898-094 101898-096
1-[5-(Aminomethyl)-2-pyridinyl]-3-pyrrolidinol dihydrochloride
1-[5-(Aminomethyl)-2-pyridinyl]-3-pyrrolidinol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇Cl₂N₃O
MW: 266.17 g/mol
MDL Number: MFCD28506179
CAS Number: 1820734-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR