[6-(4-Morpholinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052050-500MG 052050-2.5G
101898-090EA 364.62 USD
101898-090 101898-092
[6-(4-Morpholinyl)-3-pyridinyl]methanamine
[6-(4-Morpholinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD06213528
CAS Number: 771572-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR