5-(Aminomethyl)-N,N-diethyl-2-pyridinamine

Supplier: Matrix Scientific

052047-500MG 052047-2.5G
101898-078EA 364.62 USD
101898-078 101898-080
5-(Aminomethyl)-N,N-diethyl-2-pyridinamine
5-(Aminomethyl)-N,N-diethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₇N₃
MDL Number: MFCD09044973
CAS Number: 926234-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR