[6-(1-Pyrrolidinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052046-500MG 052046-2.5G
101898-074EA 364.62 USD
101898-074 101898-076
[6-(1-Pyrrolidinyl)-3-pyridinyl]methanamine
[6-(1-Pyrrolidinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MDL Number: MFCD09049100
CAS Number: 754977-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR