2-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)nicotinamide

Supplier: Matrix Scientific

052044-500MG 052044-2.5G
101898-066EA 364.62 USD
101898-066 101898-068
2-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)nicotinamide
2-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)nicotinamide

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O₂
MDL Number: MFCD13561881
CAS Number: 1220030-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR