N-Benzyl-2-chloro-N-ethylnicotinamide

Supplier: Matrix Scientific

052043-500MG 052043-2.5G
101898-062EA 364.62 USD
101898-062 101898-064
N-Benzyl-2-chloro-N-ethylnicotinamide
N-Benzyl-2-chloro-N-ethylnicotinamide

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O
MDL Number: MFCD12556690
CAS Number: 1039844-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR