(2-Chloro-3-pyridinyl)(2,3-dihydro-1H-indol-1-yl)methanone

Supplier: Matrix Scientific

052041-500MG 052041-2.5G
101898-054EA 364.62 USD
101898-054 101898-056
(2-Chloro-3-pyridinyl)(2,3-dihydro-1H-indol-1-yl)methanone
(2-Chloro-3-pyridinyl)(2,3-dihydro-1H-indol-1-yl)methanone

Hazard: Irritant

Formula: C₁₄H₁₁ClN₂O
MDL Number: MFCD01350741
CAS Number: 557781-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR