2-Chloro-N-cyclohexyl-N-ethylnicotinamide

Supplier: Matrix Scientific

052038-500MG 052038-2.5G
101898-042EA 364.62 USD
101898-042 101898-044
2-Chloro-N-cyclohexyl-N-ethylnicotinamide
2-Chloro-N-cyclohexyl-N-ethylnicotinamide

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O
MDL Number: MFCD09815619
CAS Number: 1016760-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR