N-Benzyl-2-chloro-N-methylnicotinamide

Supplier: Matrix Scientific

052037-500MG 052037-2.5G
101898-038EA 364.62 USD
101898-038 101898-040
N-Benzyl-2-chloro-N-methylnicotinamide
N-Benzyl-2-chloro-N-methylnicotinamide

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂O
MDL Number: MFCD09808525
CAS Number: 1016751-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR