2-Chloro-N-cyclohexyl-N-methylnicotinamide

Supplier: Matrix Scientific

052035-500MG 052035-2.5G
101898-030EA 364.62 USD
101898-030 101898-032
2-Chloro-N-cyclohexyl-N-methylnicotinamide
2-Chloro-N-cyclohexyl-N-methylnicotinamide

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O
MDL Number: MFCD09813284
CAS Number: 1016516-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR