(2-Chloro-3-pyridinyl)(4-hydroxy-1-piperidinyl)methanone

Supplier: Matrix Scientific

052033-500MG 052033-2.5G
101898-022EA 364.62 USD
101898-022 101898-024
(2-Chloro-3-pyridinyl)(4-hydroxy-1-piperidinyl)methanone
(2-Chloro-3-pyridinyl)(4-hydroxy-1-piperidinyl)methanone

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
MDL Number: MFCD09807894
CAS Number: 1016680-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR