(2-Chloro-3-pyridinyl)(4-methyl-1-piperidinyl)methanone

Supplier: Matrix Scientific

052028-500MG 052028-2.5G
101898-002EA 364.62 USD
101898-002 101898-004
(2-Chloro-3-pyridinyl)(4-methyl-1-piperidinyl)methanone
(2-Chloro-3-pyridinyl)(4-methyl-1-piperidinyl)methanone

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O
MDL Number: MFCD02325412
CAS Number: 353258-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR