(2-Chloro-3-pyridinyl)(3-methyl-1-piperidinyl)methanone

Supplier: Matrix Scientific

052027-500MG 052027-2.5G
101897-998EA 364.62 USD
101897-998 101898-000
(2-Chloro-3-pyridinyl)(3-methyl-1-piperidinyl)methanone
(2-Chloro-3-pyridinyl)(3-methyl-1-piperidinyl)methanone

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O
MDL Number: MFCD03385184
CAS Number: 560106-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR