(2-Chloro-3-pyridinyl)(2-methyl-1-piperidinyl)methanone
Supplier: Matrix Scientific
052026-500MG
052026-2.5G
101897-994EA
364.62
USD
101897-994
101897-996
(2-Chloro-3-pyridinyl)(2-methyl-1-piperidinyl)methanone
(2-Chloro-3-pyridinyl)(2-methyl-1-piperidinyl)methanone
Hazard: Irritant
Formula:
C₁₂H₁₅ClN₂O |
MDL Number:
MFCD01344612 CAS Number: 438613-81-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...