2-Chloro-N-ethyl-N-(2-hydroxyethyl)nicotinamide

Supplier: Matrix Scientific

052024-500MG 052024-2.5G
101897-986EA 364.62 USD
101897-986 101897-988
2-Chloro-N-ethyl-N-(2-hydroxyethyl)nicotinamide
2-Chloro-N-ethyl-N-(2-hydroxyethyl)nicotinamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD12811876
CAS Number: 1184259-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR