(2-Chloro-3-pyridinyl)(3-hydroxy-1-pyrrolidinyl)methanone
Supplier: Matrix Scientific
052022-500MG
052022-2.5G
101897-978EA
364.62
USD
101897-978
101897-980
(2-Chloro-3-pyridinyl)(3-hydroxy-1-pyrrolidinyl)methanone
(2-Chloro-3-pyridinyl)(3-hydroxy-1-pyrrolidinyl)methanone
Hazard: Irritant
Formula:
C₁₀H₁₁ClN₂O₂ |
MDL Number:
MFCD13561880 CAS Number: 1219981-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...