(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone

Supplier: Matrix Scientific

Synonyms: 4-(2-Chloropyridine-3-carbonyl)morpholine, (2-Chloropyridin-3-yl)(morpholino)methanone

052021-500MG 052021-2.5G
101897-974EA 364.62 USD
101897-974 101897-976
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂O₂
MW: 226.66 g/mol
MDL Number: MFCD00454210
CAS Number: 53062-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR