2-Chloro-N-(2-hydroxyethyl)-N-methylnicotinamide

Supplier: Matrix Scientific

052020-500MG 052020-2.5G
101897-970EA 364.62 USD
101897-970 101897-972
2-Chloro-N-(2-hydroxyethyl)-N-methylnicotinamide
2-Chloro-N-(2-hydroxyethyl)-N-methylnicotinamide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD12811677
CAS Number: 1184830-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR