N2-Methyl-N2-(tetrahydro-2H-pyran-4-ylmethyl)-2,3-pyridinediamine
Supplier: Matrix Scientific
052018-500MG
052018-2.5G
101897-962EA
364.62
USD
101897-962
101897-964
N2-Methyl-N2-(tetrahydro-2H-pyran-4-ylmethyl)-2,3-pyridinediamine
N2-Methyl-N2-(tetrahydro-2H-pyran-4-ylmethyl)-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₁₂H₁₉N₃O |
MDL Number:
MFCD13561879 CAS Number: 1219967-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...