N2-Benzyl-N2-ethyl-2,3-pyridinediamine

Supplier: Matrix Scientific

052017-500MG 052017-2.5G
101897-958EA 364.62 USD
101897-958 101897-960
N2-Benzyl-N2-ethyl-2,3-pyridinediamine
N2-Benzyl-N2-ethyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₄H₁₇N₃
MDL Number: MFCD11195376
CAS Number: 1039926-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR