2-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinamine

Supplier: Matrix Scientific

052016-500MG 052016-2.5G
101897-954EA 364.62 USD
101897-954 101897-956
2-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinamine
2-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinamine

Hazard: Irritant

Formula: C₁₄H₁₅N₃
MDL Number: MFCD08700179
CAS Number: 937603-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR