2-[4-(3-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol
Supplier: Matrix Scientific
052013-500MG
052013-2.5G
101897-942EA
364.62
USD
101897-942
101897-944
2-[4-(3-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol
2-[4-(3-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol
Hazard: Irritant
Formula:
C₁₁H₁₈N₄O |
MDL Number:
MFCD08692318 CAS Number: 1082137-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...