N2-Cyclohexyl-N2-ethyl-2,3-pyridinediamine

Supplier: Matrix Scientific

052012-500MG 052012-2.5G
101897-938EA 364.62 USD
101897-938 101897-940
N2-Cyclohexyl-N2-ethyl-2,3-pyridinediamine
N2-Cyclohexyl-N2-ethyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
MDL Number: MFCD09737899
CAS Number: 954264-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR