N2,N2-Dipropyl-2,3-pyridinediamine

Supplier: Matrix Scientific

052008-500MG 052008-2.5G
101897-922EA 364.62 USD
101897-922 101897-924
N2,N2-Dipropyl-2,3-pyridinediamine
N2,N2-Dipropyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₉N₃
MDL Number: MFCD09045871
CAS Number: 926240-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR