1-(3-Amino-2-pyridinyl)-4-piperidinol

Supplier: Matrix Scientific

052007-500MG 052007-2.5G
101897-918EA 364.62 USD
101897-918 101897-920
1-(3-Amino-2-pyridinyl)-4-piperidinol
1-(3-Amino-2-pyridinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD09742956
CAS Number: 954570-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR