2-(1-Azepanyl)-3-pyridinamine

Supplier: Matrix Scientific

052005-500MG 052005-2.5G
101897-910EA 364.62 USD
101897-910 101897-912
2-(1-Azepanyl)-3-pyridinamine
2-(1-Azepanyl)-3-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD08700177
CAS Number: 94520-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR