2-(4-Benzyl-1-piperazinyl)-3-pyridinylamine

Supplier: Matrix Scientific

052004-500MG 052004-2.5G
101897-906EA 364.62 USD
101897-906 101897-908
2-(4-Benzyl-1-piperazinyl)-3-pyridinylamine
2-(4-Benzyl-1-piperazinyl)-3-pyridinylamine

Hazard: Irritant

Formula: C₁₆H₂₀N₄
MDL Number: MFCD08692316
CAS Number: 937603-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR