2-(2-Methyl-1-piperidinyl)-3-pyridinylamine
Supplier: Matrix Scientific
052001-500MG
052001-2.5G
101897-894EA
364.62
USD
101897-894
101897-896
2-(2-Methyl-1-piperidinyl)-3-pyridinylamine
2-(2-Methyl-1-piperidinyl)-3-pyridinylamine
Hazard: Irritant
Formula:
C₁₁H₁₇N₃ |
MDL Number:
MFCD09043299 CAS Number: 926200-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...