N2,N2-Diallyl-2,3-pyridinediamine

Supplier: Matrix Scientific

052000-500MG 052000-2.5G
101897-890EA 364.62 USD
101897-890 101897-892
N2,N2-Diallyl-2,3-pyridinediamine
N2,N2-Diallyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₅N₃
MDL Number: MFCD11562396
CAS Number: 1082177-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR