N2-Butyl-N2-methyl-2,3-pyridinediamine

Supplier: Matrix Scientific

051998-500MG 051998-2.5G
101897-882EA 364.62 USD
101897-882 101897-884
N2-Butyl-N2-methyl-2,3-pyridinediamine
N2-Butyl-N2-methyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₀H₁₇N₃
MDL Number: MFCD09812485
CAS Number: 1016682-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR