1-(3-Amino-2-pyridinyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051997-500MG 051997-2.5G
101897-878EA 364.62 USD
101897-878 101897-880
1-(3-Amino-2-pyridinyl)-3-pyrrolidinol
1-(3-Amino-2-pyridinyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₉H₁₃N₃O
MW: 179.22 g/mol
MDL Number: MFCD13561878
CAS Number: 1220028-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR