2-(1-Piperidinyl)-3-pyridinamine

Supplier: Matrix Scientific

051996-500MG 051996-2.5G
101897-874EA 461.54 USD
101897-874 101897-876
2-(1-Piperidinyl)-3-pyridinamine
2-(1-Piperidinyl)-3-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MDL Number: MFCD08700153
CAS Number: 5028-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR