2-[(3-Amino-2-pyridinyl)(methyl)amino]-1-ethanol

Supplier: Matrix Scientific

051995-500MG 051995-2.5G
101897-870EA 364.62 USD
101897-870 101897-872
2-[(3-Amino-2-pyridinyl)(methyl)amino]-1-ethanol
2-[(3-Amino-2-pyridinyl)(methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₈H₁₃N₃O
MDL Number: MFCD11562401
CAS Number: 1082137-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR