N2,N2-Diethyl-2,3-pyridinediamine

Supplier: Matrix Scientific

051994-500MG 051994-2.5G
101897-866EA 364.62 USD
101897-866 101897-868
N2,N2-Diethyl-2,3-pyridinediamine
N2,N2-Diethyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₉H₁₅N₃
MDL Number: MFCD09045566
CAS Number: 926226-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR