2-(1-Pyrrolidinyl)-3-pyridinamine

Supplier: Matrix Scientific

051993-500MG 051993-2.5G
101897-862EA 307.69 USD
101897-862 101897-864
2-(1-Pyrrolidinyl)-3-pyridinamine
2-(1-Pyrrolidinyl)-3-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃N₃
MW: 163.22 g/mol
MDL Number: MFCD08700176
CAS Number: 5028-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR