N2,N2-Dimethyl-2,3-pyridinediamine

Supplier: Matrix Scientific

051992-500MG 051992-2.5G
101897-858EA 384.62 USD
101897-858 101897-860
N2,N2-Dimethyl-2,3-pyridinediamine
N2,N2-Dimethyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₇H₁₁N₃
MDL Number: MFCD09737533
CAS Number: 5028-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR