2-(2,3-Dihydro-1H-indol-1-yl)nicotinic acid
Supplier: Matrix Scientific
051988-500MG
051988-2.5G
101897-842EA
166.15
USD
101897-842
101897-844
2-(2,3-Dihydro-1H-indol-1-yl)nicotinic acid
2-(2,3-Dihydro-1H-indol-1-yl)nicotinic acid
Hazard: Irritant
Formula:
C₁₄H₁₂N₂O₂ |
MDL Number:
MFCD09896574 CAS Number: 1017138-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...