2-[4-(2-Hydroxyethyl)-1-piperazinyl]nicotinic acid

Supplier: Matrix Scientific

051986-500MG 051986-2.5G
101897-834EA 364.62 USD
101897-834 101897-836
2-[4-(2-Hydroxyethyl)-1-piperazinyl]nicotinic acid
2-[4-(2-Hydroxyethyl)-1-piperazinyl]nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃
MDL Number: MFCD11107299
CAS Number: 111855-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR