2-[Cyclohexyl(ethyl)amino]nicotinic acid
Supplier: Matrix Scientific
051985-500MG
051985-2.5G
101897-830EA
364.62
USD
101897-830
101897-832
2-[Cyclohexyl(ethyl)amino]nicotinic acid
2-[Cyclohexyl(ethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₄H₂₀N₂O₂ |
MDL Number:
MFCD07379604 CAS Number: 1019350-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...