2-[Benzyl(methyl)amino]nicotinic acid

Supplier: Matrix Scientific

051984-500MG 051984-2.5G
101897-826EA 364.62 USD
101897-826 101897-828
2-[Benzyl(methyl)amino]nicotinic acid
2-[Benzyl(methyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD07379597
CAS Number: 35174-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR