2-(2-Ethyl-1-piperidinyl)nicotinic acid

Supplier: Matrix Scientific

051983-500MG 051983-2.5G
101897-822EA 364.62 USD
101897-822 101897-824
2-(2-Ethyl-1-piperidinyl)nicotinic acid
2-(2-Ethyl-1-piperidinyl)nicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD07379605
CAS Number: 1094523-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR