2-(3-Oxo-1-piperazinyl)nicotinic acid

Supplier: Matrix Scientific

051980-500MG 051980-2.5G
101897-810EA 364.62 USD
101897-810 101897-812
2-(3-Oxo-1-piperazinyl)nicotinic acid
2-(3-Oxo-1-piperazinyl)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₃
MDL Number: MFCD11132187
CAS Number: 1019359-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR