2-(1-Azepanyl)nicotinic acid

Supplier: Matrix Scientific

051979-500MG 051979-2.5G
101897-806EA 364.62 USD
101897-806 101897-808
2-(1-Azepanyl)nicotinic acid
2-(1-Azepanyl)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD03822745
CAS Number: 571913-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR