2-(4-Benzyl-1-piperazinyl)nicotinic acid
Supplier: Matrix Scientific
051978-500MG
051978-2.5G
101897-802EA
364.62
USD
101897-802
101897-804
2-(4-Benzyl-1-piperazinyl)nicotinic acid
2-(4-Benzyl-1-piperazinyl)nicotinic acid
Hazard: Irritant
Formula:
C₁₇H₁₉N₃O₂ |
MDL Number:
MFCD07379598 CAS Number: 247117-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...