2-(4-Methyl-1-piperidinyl)nicotinic acid

Supplier: Matrix Scientific

051976-500MG 051976-2.5G
101897-794EA 364.62 USD
101897-794 101897-796
2-(4-Methyl-1-piperidinyl)nicotinic acid
2-(4-Methyl-1-piperidinyl)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD03114516
CAS Number: 460363-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR