2-(2-Methyl-1-piperidinyl)nicotinic acid
Supplier: Matrix Scientific
051974-500MG
051974-2.5G
101897-786EA
364.62
USD
101897-786
101897-788
2-(2-Methyl-1-piperidinyl)nicotinic acid
2-(2-Methyl-1-piperidinyl)nicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD11131980 CAS Number: 1019461-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...