2-(Diallylamino)nicotinic acid

Supplier: Matrix Scientific

051973-500MG 051973-2.5G
101897-782EA 364.62 USD
101897-782 101897-784
2-(Diallylamino)nicotinic acid
2-(Diallylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD11131941
CAS Number: 1019461-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR