2-[Butyl(methyl)amino]nicotinic acid

Supplier: Matrix Scientific

051971-500MG 051971-2.5G
101897-774EA 364.62 USD
101897-774 101897-776
2-[Butyl(methyl)amino]nicotinic acid
2-[Butyl(methyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD11132104
CAS Number: 1019465-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR